Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Quickfit
®
three-neck round-bottom flasks with angled necks
Products
Product Category
flasks (11)
Manufacturer
Capacity (mL)
Height (mm)
Closure Type
Package Size
Sterility
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
Z303534
Quickfit
®
three-neck round-bottom flasks with angled necks,
capacity 250 mL, center joint: ST/NS 24/29, side joint: ST/NS 19/26
Expand
View Pricing
Z334510
Quickfit
®
three-neck round-bottom flasks with angled necks,
center joint: ST/NS 14/23, side joint: ST/NS 14/23
Expand
View Pricing
Z303577
Quickfit
®
three-neck round-bottom flasks with angled necks,
capacity 1,000 mL, center joint: ST/NS 24/29, side joint: ST/NS 19/26
Expand
View Pricing
Z303542
Quickfit
®
three-neck round-bottom flasks with angled necks,
capacity 250 mL, center joint: ST/NS 29/32, side joint: ST/NS 19/26
Expand
View Pricing
Z334545
Quickfit
®
three-neck round-bottom flasks with angled necks,
capacity 50 mL, center joint: ST/NS 19/26, side joint: ST/NS 14/23
Expand
View Pricing
Z303569
Quickfit
®
three-neck round-bottom flasks with angled necks,
capacity 500 mL, center joint: ST/NS 29/32, side joint: ST/NS 19/26
Expand
View Pricing
Z309257
Quickfit
®
three-neck round-bottom flasks with angled necks,
capacity 100 mL, center joint: ST/NS 24/29, side joint: ST/NS 14/23
Expand
View Pricing
Z334537
Quickfit
®
three-neck round-bottom flasks with angled necks,
capacity 50 mL, center joint: ST/NS 14/23, side joint: ST/NS 14/23
Expand
View Pricing
Z303518
Quickfit
®
three-neck round-bottom flasks with angled necks,
capacity 100 mL, center joint: ST/NS 24/29, side joint: ST/NS 19/26
Expand
View Pricing
Z303526
Quickfit
®
three-neck round-bottom flasks with angled necks,
capacity 250 mL, center joint: ST/NS 19/26, side joint: ST/NS 19/26
Expand
View Pricing
Z303496
Quickfit
®
three-neck round-bottom flasks with angled necks,
capacity 100 mL, center joint: ST/NS 19/26, side joint: ST/NS 14/23
Expand
View Pricing