
DCA Deblock (0.36M dichloroacetic acid in toluene)

Synonym(s):
DCA Deblock
Empirical Formula (Hill Notation):
C2H2Cl2O2
CAS Number:
Molecular Weight:
128.94
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing