Skip to Content
Merck
All Photos(1)

Key Documents

707066

Sigma-Aldrich

DCA Deblock (0.36M dichloroacetic acid in toluene)

Synonym(s):

DCA Deblock, DEBLOCK (0.36M dichloroacetic acid in toluene)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H2Cl2O2
CAS Number:
Molecular Weight:
128.94
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

form

liquid

Quality Level

concentration

≤100 ppm in H2O

bp

110-111 °C

density

0.884 g/mL at 25 °C

SMILES string

OC(=O)C(Cl)Cl

InChI

1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)

InChI key

JXTHNDFMNIQAHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Deblock solution, containing dichloroacetic acid (DCA) in dichloromethane or Toluene, removes the dimethoxytrityl (DMT) protecting the group from the 5′ hydroxyl moiety of nucleotides already incorporated into the growing nucleic acid, prior to the addition of the next phosphoramidite. Removal of the DMT allows the unprotected 5′ hydroxyl moiety to react with a new phosphoramidite in a subsequent extension reaction.

Application

DCA Deblock (0.36M dichloroacetic acid in toluene) is suitable for the removal of 4,4Adimethoxytrityl (DMTr) protecting groups for the synthesis of antisense oligonucleotides in high yield and purity.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Lact. - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.0 °F

Flash Point(C)

5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A H Krotz et al.
Bioorganic & medicinal chemistry, 7(3), 435-439 (1999-04-29)
It is demonstrated that a solution of dichloroacetic acid in toluene removes dimethoxytrityl groups from the 5'-terminus of an antisense phosphorothioate oligodeoxyribonucleotide (ISIS 5132/CGP69846A) during synthesis on solid support cleanly and efficiently. It is therefore suggested to replace health hazardous
Synthesis of an antisense oligonucleotide targeted against C-raf kinase: efficient oligonucleotide synthesis without chlorinated solvents
A H Krotz and D L Cole V T Ravikumar
Bioorganic & Medicinal Chemistry, 7(3), 435-439 (1999)
Haihua Xiao et al.
Chemical communications (Cambridge, England), 48(87), 10730-10732 (2012-09-27)
A multifunctional hybrid platinum(IV) prodrug, which consists of both the mitochondria-targeting drug DCA and the DNA-crosslinking drug cisplatin, was synthesized and tethered to a carrier polymer to further self-assemble into micelles for intracellular delivery.
Toshimitsu Ohashi et al.
International journal of cancer, 133(5), 1107-1118 (2013-02-20)
The activation of oncogenic signaling pathways induces the reprogramming of glucose metabolism in tumor cells and increases lactic acid secretion into the tumor microenvironment. This is a well-known characteristic of tumor cells, termed the Warburg effect, and is a candidate
Jae Mo Park et al.
Neuro-oncology, 15(4), 433-441 (2013-01-19)
The metabolic phenotype that derives disproportionate energy via glycolysis in solid tumors, including glioma, leads to elevated lactate labeling in metabolic imaging using hyperpolarized [1-(13)C]pyruvate. Although the pyruvate dehydrogenase (PDH)-mediated flux from pyruvate to acetyl coenzyme A can be indirectly

Articles

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

Fluorescence lifetime measurement is advantageous over intensity-based measurements. Applications include fluorescence lifetime assays, sensing and FLI.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service