Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Büchi
®
evaporating flask, pear-shaped
Products
Product Category
flasks (11)
Brand
Sigma-Aldrich (7)
Manufacturer
Capacity (mL)
Volume (ml)
Closure Type
Package Size
Sterility
Style
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
Z403016
Büchi
®
evaporating flask, pear-shaped,
volume 2,000 mL, glass, joint: ST/NS 24/40
Expand
View Pricing
Z319449
Büchi
®
evaporating flask, pear-shaped,
glass, volume 250 mL, joint: ST/NS 29/32
Expand
View Pricing
Z402990
Büchi
®
evaporating flask, pear-shaped,
volume 1,000 mL, glass, joint: ST/NS 24/40
Expand
View Pricing
Z403008
Büchi
®
evaporating flask, pear-shaped,
volume 1,000 mL, plastic-coated glass, joint: ST/NS 24/40
Expand
View Pricing
Z403032
Büchi
®
evaporating flask, pear-shaped,
volume 3,000 mL, glass, joint: ST/NS 24/40
Expand
View Pricing
Z402982
Büchi
®
evaporating flask, pear-shaped,
volume 500 mL, glass, joint: ST/NS 24/40
Expand
View Pricing
Z402958
Büchi
®
evaporating flask, pear-shaped,
volume 50 mL, glass, joint: ST/NS 24/40
Expand
View Pricing
Z402974
Büchi
®
evaporating flask, pear-shaped,
volume 250 mL, glass, joint: ST/NS 24/40
Expand
View Pricing
Z402966
Büchi
®
evaporating flask, pear-shaped,
volume 100 mL, glass, joint: ST/NS 24/40
Expand
View Pricing
Z403024
Büchi
®
evaporating flask, pear-shaped,
volume 2,000 mL, plastic-coated glass, joint: ST/NS 24/40
Expand
View Pricing
Z403040
Büchi
®
evaporating flask, pear-shaped,
volume 3,000 mL, plastic-coated glass, joint: ST/NS 24/40
Expand
View Pricing