Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap
Products
Product Category
bottles (15)
Manufacturer
Capacity (mL)
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
DWKBWP1000P
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap,
round translucent polypropylene bottle, capacity 1000 mL
Expand
View Pricing
DWKBWP0250P
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap,
round translucent polypropylene bottle, capacity 250 mL
Expand
View Pricing
DWKBWP0500P
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap,
round translucent polypropylene bottle, capacity 500 mL
Expand
View Pricing
DWKBWP0060AP
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap,
round amber polypropylene bottle, capacity 60 mL
Expand
View Pricing
DWKBLP0100P
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap,
round translucent polypropylene bottle, capacity 100 mL
Expand
View Pricing
DWKBWP1000AP
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap,
round amber polypropylene bottle, capacity 1000 mL
Expand
View Pricing
DWKBWP2000P
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap,
round translucent polypropylene bottle, capacity 2000 mL
Expand
View Pricing
DWKBWP0500AP
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap,
round amber polypropylene bottle, capacity 500 mL
Expand
View Pricing
DWKBLP0150P
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap,
round translucent polypropylene bottle, capacity 150 mL
Expand
View Pricing
DWKBWP0150AP
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap,
round amber polypropylene bottle, capacity 150 mL
Expand
View Pricing
DWKBWP0250AP
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap,
round amber polypropylene bottle, capacity 250 mL
Expand
View Pricing
DWKBWP0060PN
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap,
round translucent polypropylene bottle, capacity 60 mL
Expand
View Pricing
DWKBLP0125P
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap,
round translucent polypropylene bottle, capacity 1250 mL
Expand
View Pricing
DWKBWP2000AP
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap,
round amber polypropylene bottle, capacity 2000 mL
Expand
View Pricing
DWKBWP0030PN
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap,
round translucent polypropylene bottle, capacity 30 mL
Expand
View Pricing