Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
α-Glycerophosphate Dehydrogenase from rabbit muscle
Synonym(s):
sn
-Glycerol-3-phosphate Dehydrogenase from rabbit muscle,
sn
-Glycerol-3-phosphate:NAD
+
2-oxidoreductase
CAS Number:
9075-65-4
EC Number:
232-982-3
Products
Brand
Sigma-Aldrich (2)
Biological Source
Color
Physical Form
Tissue
Chemical Composition
Quality Segment
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
G6880
α-Glycerophosphate Dehydrogenase from rabbit muscle,
Type X, lyophilized powder, ≥100 units/mg protein
Expand
View Pricing
G6751
α-Glycerophosphate Dehydrogenase from rabbit muscle,
Type I, ammonium sulfate suspension, 100-300 units/mg protein
Expand
View Pricing