Skip to Content
Merck
All Photos(1)

Documents

G6880

Sigma-Aldrich

α-Glycerophosphate Dehydrogenase from rabbit muscle

Type X, lyophilized powder, ≥100 units/mg protein

Synonym(s):

sn-Glycerol-3-phosphate Dehydrogenase from rabbit muscle, sn-Glycerol-3-phosphate:NAD+ 2-oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type X

Quality Level

form

lyophilized powder

specific activity

≥100 units/mg protein

composition

Protein, ≥75%

foreign activity

Lactic dehydrogenase, pyruvate kinase, aldolase, and glyceraldehyde-3-phosphate dehydrogenase ≤0.01%
Triosephosphate isomerase ≤0.02%

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

α-glycerophosphate dehydrogenase was used in 2-deoxy-ribose 5-phosphate aldolase (DERA) cleavage (retroaldol) assay.

Biochem/physiol Actions

α-glycerophosphate dehydrogenase catalyzes the conversion of dihydroxyacetone to glycerol phosphate.

Unit Definition

One unit will convert 1.0 μmole of dihydroxyacetone phosphate to α-glycerophosphate per min at pH 7.4 at 25 °C.

Physical form

Lyophilized sulfate-free powder containing buffer salts as citrate and EDTA

Analysis Note

Protein determined by biuret

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grace DeSantis et al.
Bioorganic & medicinal chemistry, 11(1), 43-52 (2002-12-07)
2-Deoxyribose-5-phosphate aldolase (DERA, EC 4.1.2.4) catalyzes the reversible aldol reaction between acetaldehyde and D-glyceraldehyde-3-phosphate to generate D-2-deoxyribose-5-phosphate. It is unique among the aldolases as it catalyzes the reversible asymmetric aldol addition reaction of two aldehydes. In order to expand the
Yongxiu Fan et al.
Parasitology research, 109(3), 621-626 (2011-03-17)
Glycerol 3-phosphate dehydrogenase (GPD) plays an important role in the energy metabolism and nutrition metabolism. In order to know about the biological functions of GPD of Clonorchis sinensis (C. sinensis), we identified a complete gene coding GPD from C. sinensis
Yunzhen Gao et al.
Journal of applied genetics, 52(4), 451-458 (2011-05-18)
Soluble glycerol-3-phosphate dehydrogenase 1 (GPD1, EC 1.1.1.8) plays important roles in the synthesis of triacylglycerol and in the glycerol-3-phosphate shutter. Though GPD1 is expressed in most adult tissues, little is known about the regulation of its expression. In this study
Peter M Fernandes et al.
The Biochemical journal, 477(22), 4425-4441 (2020-11-04)
6-Phosphofructokinase-1-kinase (PFK) tetramers catalyse the phosphorylation of fructose 6-phosphate (F6P) to fructose 1,6-bisphosphate (F16BP). Vertebrates have three PFK isoforms (PFK-M, PFK-L, and PFK-P). This study is the first to compare the kinetics, structures, and transcript levels of recombinant human PFK
Camila Guindalini et al.
Experimental biology and medicine (Maywood, N.J.), 235(1), 52-56 (2010-04-21)
Glycerol-3-phosphate dehydrogenase 1 (GPD1) is considered to be a key enzyme that connects carbohydrate and lipid metabolism. This gene is induced in response to sleep deprivation, suggesting a potential role for this enzyme in the manifestation of obstructive sleep apnea

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service