
8arm-PEG20K

Synonym(s):
8arm-PEG-Norbornene, tripentaerythritol core, 8arm-PEG-Maleimide, 8arm-PEG-NH2, tripentaerythritol core, 8arm-PEG-NH2, hexaglycerol core, 8arm-PEG-Vinylsulfone, tripentaerythritol core, 8arm-PEG-Acrylate, hexaglycerol core
Linear Formula:
R(O(CH2CH2O)nCOCH2CH2CH2COOC4H4O2N)8
Products
Brand
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Showing 1-7 of 7
Compare
Product Number
Description
Pricing