JKA8022
8arm-PEG20K
average MN 20,000, Succinimidyl Glutarate, hexaglycerol core
Synonym(s):
Polyethylene glycol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
R(O(CH2CH2O)nCOCH2CH2CH2COOC4H4O2N)8
UNSPSC Code:
51171641
NACRES:
NA.23
Recommended Products
Product Name
8arm-PEG20K-Succinimidyl Glutarate, Hexaglycerol core, average Mn 20000
form
solid
mol wt
average Mn 20000
storage temp.
−20°C
Related Categories
Application
Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization
Legal Information
Product of JenKem Technology
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tong, X. and F. Yang
Biomaterials, 35(6) (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service