![4-Methylpyrazole hydrochloride alcohol dehydrogenase inhibitor](/deepweb/assets/sigmaaldrich/product/structures/138/547/53ca07ff-489b-483c-beeb-52ff0b74ec51/640/53ca07ff-489b-483c-beeb-52ff0b74ec51.png)
4-Methylpyrazole hydrochloride
![4-Methylpyrazole hydrochloride alcohol dehydrogenase inhibitor](/deepweb/assets/sigmaaldrich/product/structures/138/547/53ca07ff-489b-483c-beeb-52ff0b74ec51/640/53ca07ff-489b-483c-beeb-52ff0b74ec51.png)
Synonym(s):
Fomepizole
Empirical Formula (Hill Notation):
C4H6N2 · HCl
CAS Number:
Molecular Weight:
118.56
Products
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing