Skip to Content
Merck

1672600

USP

Tramadol hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(±)-cis-2-(Dimethylaminomethyl)-1-(3-methoxyphenyl)cyclohexanol hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H25NO2 · HCl
CAS Number:
Molecular Weight:
299.84
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

tramadol

manufacturer/tradename

USP

drug control

USDEA Schedule IV

application(s)

pharmaceutical (small molecule)
pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

Cl.COc1cccc(c1)[C@@]2(O)CCCC[C@@H]2CN(C)C

InChI

1S/C16H25NO2.ClH/c1-17(2)12-14-7-4-5-10-16(14,18)13-8-6-9-15(11-13)19-3;/h6,8-9,11,14,18H,4-5,7,10,12H2,1-3H3;1H/t14-,16+;/m1./s1

InChI key

PPKXEPBICJTCRU-XMZRARIVSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tramadol hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Tramadol Hydrochloride Tablets
  • Tramadol Hydrochloride and Acetaminophen Tablets
  • Tramadol Hydrochloride Extended-Release Tablets
  • Tramadol Hydrochloride Compounded Oral Suspension
  • Tramadol Hydrochloride Compounded Oral Suspension, Veterinary
  • Tramadol Hydrochloride and Acetaminophen Compounded Oral Suspension

Biochem/physiol Actions

Opioid analgesic; P450 substrate.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Tramadol hydrochloride


Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library