Skip to Content
Merck

Skip To

1653117

USP

Theophylline melting point standard

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Theophylline, 1,3-Dimethylxanthine, 2,6-Dihydroxy-1,3-dimethylpurine, 3,7-Dihydro-1,3-dimethyl-1H-purine-2,6-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C7H8N4O2
CAS Number:
Molecular Weight:
180.16
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
13463
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

theophylline

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1C(=O)N(C)c2[nH]cnc2C1=O

InChI

1S/C7H8N4O2/c1-10-5-4(8-3-9-5)6(12)11(2)7(10)13/h3H,1-2H3,(H,8,9)

InChI key

ZFXYFBGIUFBOJW-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Theophylline melting point standard USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Phosphodiesterase inhibitor; diuretic; cardiac stimulant; muscle relaxant; asthma medication.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BP327PHR10231653004
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

BP

manufacturer/tradename

-

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

food and beverages
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-30°C

storage temp.

-

Gene Information

human ... ADORA1(134), ADORA2A(135), ADORA2B(136), ADORA3(140), PDE3A(5139), PDE3B(5140), PDE4A(5141), PDE4B(5142), PDE4C(5143), PDE4D(5144)

Gene Information

human ... ADORA1(134), ADORA2A(135), ADORA2B(136), ADORA3(140), PDE3A(5139), PDE3B(5140), PDE4A(5141), PDE4B(5142), PDE4C(5143), PDE4D(5144)

Gene Information

human ... ADORA1(134), ADORA2A(135), ADORA2B(136), ADORA3(140), PDE3A(5139), PDE3B(5140), PDE4A(5141), PDE4B(5142), PDE4C(5143), PDE4D(5144)

Gene Information

human ... ADORA1(134), ADORA2A(135), ADORA2B(136), ADORA3(140), PDE3A(5139), PDE3B(5140), PDE4A(5141), PDE4B(5142), PDE4C(5143), PDE4D(5144)


Still not finding the right product?


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters