Skip to Content
Merck

Skip To

1629000

USP

Sulfamethazine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4,6-Dimethylsulfadiazine, 4-Amino-N-(4,6-dimethyl-2-pyrimidinyl)benzenesulfonamide, Sulfadimethyldiazine, Sulfadimidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H14N4O2S
CAS Number:
Molecular Weight:
278.33
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
261304
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

sulfamethazine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cc1cc(C)nc(NS(=O)(=O)c2ccc(N)cc2)n1

InChI

1S/C12H14N4O2S/c1-8-7-9(2)15-12(14-8)16-19(17,18)11-5-3-10(13)4-6-11/h3-7H,13H2,1-2H3,(H,14,15,16)

InChI key

ASWVTGNCAZCNNR-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfamethazine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monograph such as Sulfamethazine Granulated.

Biochem/physiol Actions

An antimicrobial sulfur drug. Induces CYP3A4 expression and acetylated by N-acetyltransferase. Exhibits sex dependent pharmacokinetics, metabolized by the male specific isoform CYP2C11.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR3136Y0001495S6256
grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

EDQM

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

application(s)

-

format

neat

format

-

format

neat

format

-

API family

sulfamethazine

API family

sulfamethazine

API family

sulfadimidine

API family

-


Still not finding the right product?

Explore all of our products under Sulfamethazine


Storage Class

11 - Combustible Solids

wgk

WGK 2

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters