Skip to Content
Merck

1605205

USP

Ropinirole hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-[2-(Dipropylamino)ethyl]-1,3-dihydro-2H-indol-2-one hydrochloride, SKF 101468 hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H24N2O · HCl
CAS Number:
Molecular Weight:
296.84
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

ropinirole

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CCCN(CCC)CCc1cccc2NC(=O)Cc12

InChI

1S/C16H24N2O.ClH/c1-3-9-18(10-4-2)11-8-13-6-5-7-15-14(13)12-16(19)17-15;/h5-7H,3-4,8-12H2,1-2H3,(H,17,19);1H

InChI key

XDXHAEQXIBQUEZ-UHFFFAOYSA-N

Gene Information

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ropinirole hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Ropinirole Extended-Release Tablets
  • Ropinirole Hydrochloride
  • Ropinirole Tablets

Biochem/physiol Actions

An agonist at the D2 and D3 dopamine receptor subtypes, binding with higher affinity to D3 than to D2 or D4. It has negligible effect on D1-receptors. It has medium in vitro affinity to opioid receptors. Ropinirole is said to have virtually no affinity to 5-HT1, 5-HT2, benzodiazepine, GABA, muscarinic, α1-, α2-, and β-adrenoreceptors. Used as antiparkinsonian drug.
Selective D2 dopamine receptor agonist, antiparkinsonian

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Ropinirole hydrochloride


pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library