Skip to Content
Merck
All Photos(1)

Key Documents

1602003

USP

Resorcinol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1,3-Benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
906905
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

3.8 (vs air)

vapor pressure

1 mmHg ( 21.1 °C)

API family

resorcinol

autoignition temp.

1126 °F

manufacturer/tradename

USP

bp

178 °C/16 mmHg (lit.)

mp

109-112 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Oc1cccc(O)c1

InChI

1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H

InChI key

GHMLBKRAJCXXBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Resorcinol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Fluorescein Injection
  • Fluorescein Sodium
  • Resorcinol
  • Resorcinol and Sulfur Topical Suspension

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 1 Oral - STOT SE 2 Oral

Target Organs

Central nervous system,Blood, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M N Kardar et al.
Phytochemistry, 106, 156-163 (2014-08-12)
Chemical investigation of a sample of propolis originating from North-Western Cameroon led to the isolation of thirteen alk(en)ylphenols (1-13) (inseparable mixture) along with α-amyrin (14), β-amyrin (15), lupeol (16), cycloartenol (17), mangiferonic acid (18), ambonic acid (19), mangiferolic acid (20)
Sebastian Weis et al.
Pancreas, 43(8), 1277-1285 (2014-08-08)
The gene p8 was initially described in pancreatic tissue during acute experimental pancreatitis, a disease that is characterized by a systemic immune response. Although early reports suggested that p8 affects leukocyte migration during acute pancreatitis (AP), no studies revealing its
Claudio B S Oliveira et al.
Molecules (Basel, Switzerland), 19(5), 5898-5912 (2014-05-09)
A resorcinarene derivative of vanillin, resvan, was synthesized and characterized by spectroscopic techniques. We measured the cytotoxicity (in vivo and in vitro), antioxidant and anti-Toxoplasma activities of vanillin and the resorcinarene compound. Here we show that vanillin has a dose-dependent
Valerio Chiurchiù et al.
International immunology, 27(3), 153-160 (2014-10-27)
G protein-coupled receptor 55 (GPR55) is activated by endogenous, plant-derived and synthetic cannabinoids. Recent studies reported a broad tissue distribution for GPR55 and found prominent roles for this receptor in inflammatory pain, gut and bone physiology, as well as cancer.
Linlin Qi et al.
Analytical and bioanalytical chemistry, 407(13), 3617-3625 (2015-03-23)
An aqueous two-phase microfluidics (ATPM) method suitable for selective extraction of bisphenol A (BPA) in aqueous samples was developed, and a functional ionic liquid of N, N, N-trioctyl ammonium propionate (TOAP) was specially employed for the formation of a parallel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service