Skip to Content
Merck

Skip To

1568007

USP

Progesterone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Pregnene-3,20-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1915950
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

progesterone

manufacturer/tradename

USP

mp

128-132 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19H,4-11H2,1-3H3/t16-,17+,18-,19-,20-,21+/m0/s1

InChI key

RJKFOVLPORLFTN-LEKSSAKUSA-N

Gene Information

human ... PGR(5241)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Progesterone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Progesterone Compounded Vaginal Inserts
  • Progesterone Injection
  • Progesterone Intrauterine Contraceptive System

Biochem/physiol Actions

Induces maturation and secretory activity of the uterine endothelium; suppresses ovulation.
Induces maturation and secretory activity of the uterine endothelium; suppresses ovulation. Progesterone is implicated in the etiology of breast cancer.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
Steroid hormone produced by the corpus luteum.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BP449PHR1142P3972
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

manufacturer/tradename

USP

manufacturer/tradename

BP

manufacturer/tradename

-

manufacturer/tradename

-

format

neat

format

neat

format

neat

format

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

Gene Information

human ... PGR(5241)

Gene Information

human ... PGR(5241)

Gene Information

human ... PGR(5241)

Gene Information

human ... ABCB1(5243), AR(367), CYP2C9(1559), EBP(10682), ESR1(2099), ESR2(2100), NR3C1(2908), NR3C2(4306), PGR(5241), SERPINA6(866)
mouse ... Esr1(13982)
rat ... Ar(24208), Nr3c1(24413), Pgr(25154)

API family

progesterone

API family

progesterone

API family

progesterone

API family

-


Still not finding the right product?

Explore all of our products under Progesterone


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters