Select a Size
About This Item
grade
pharmaceutical primary standard
manufacturer/tradename
USP
mp
218-220 °C (lit.)
application(s)
USP Biologics
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
CC(C)(C)c1cc(CN2C(=O)N(Cc3cc(c(O)c(c3)C(C)(C)C)C(C)(C)C)C(=O)N(Cc4cc(c(O)c(c4)C(C)(C)C)C(C)(C)C)C2=O)cc(c1O)C(C)(C)C
InChI
1S/C48H69N3O6/c1-43(2,3)31-19-28(20-32(37(31)52)44(4,5)6)25-49-40(55)50(26-29-21-33(45(7,8)9)38(53)34(22-29)46(10,11)12)42(57)51(41(49)56)27-30-23-35(47(13,14)15)39(54)36(24-30)48(16,17)18/h19-24,52-54H,25-27H2,1-18H3
InChI key
VNQNXQYZMPJLQX-UHFFFAOYSA-N
General description
Plastic additive 6 is crucial in enhancing the flexibility and durability of polymeric materials in biologic drug packaging. By ensuring that packaging remains robust and resistant to degradation, this additive helps maintain the efficacy and safety of biologic products. As a USP reference standard, its role is to support consistent, high-quality packaging for biologics, meeting strict regulatory guidelines.
The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
Still not finding the right product?
Explore all of our products under Plastic additive 6
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1544971-200MG | 04061833832448 |