Skip to Content
Merck
All Photos(1)

Documents

1530503

USP

L-Phenylalanine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(S)-2-Amino-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
165.19
Beilstein:
1910408
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

phenylalanine

manufacturer/tradename

USP

mp

270-275 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N[C@@H](Cc1ccccc1)C(O)=O

InChI

1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1

InChI key

COLNVLDHVKWLRT-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

L-Phenylalanine USP reference standard suitable for use in specified USP compendial quality tests and assays.

Also used to prepare internal standard, standard, Sample, and system suitability solution during the assay and impurity analysis by using liquid chromatography coupled with UV detector according to the given below monographs of United States Pharmacopeia (USP): 
  • Acetylcysteine Solution
  • Acetylcysteine
  • Phenylalanine  
  • Glutathione

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phenylalanine
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 3509-3509 (2023)
Glutathione
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(3), 5054-5054 (2023)
Acetylcysteine Solution
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 49(2), 82-82 (2020)
Acetylcysteine
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 49(2), 82-82 (2020)
G Neurauter et al.
Current drug metabolism, 9(7), 622-627 (2008-09-11)
Neuropsychiatric symptoms like mood changes and depression are common in patients with chronic inflammatory disorders such as infections, autoimmune diseases or cancer. The pathogenesis of these symptoms is still unclear. Pro-inflammatory stimuli interfere not only with the neural circuits and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service