Skip to Content
Merck

1494920

USP

Pantoprazole Related Compound C

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

5-(Difluoromethoxy)-2-mercapto-1H-benzimidazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H6F2N2OS
CAS Number:
Molecular Weight:
216.21
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

pantoprazole

manufacturer/tradename

USP

mp

239-243 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

FC(F)Oc1ccc2[nH]c(S)nc2c1

InChI

1S/C8H6F2N2OS/c9-7(10)13-4-1-2-5-6(3-4)12-8(14)11-5/h1-3,7H,(H2,11,12,14)

InChI key

HJMVPNAZPFZXCP-UHFFFAOYSA-N

Application

Pantoprazole Related Compound C USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monograph such as Pantoprazole Sodium

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Pantoprazole Related Compound C


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library