Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C17H20N4OS
CAS Number:
Molecular Weight:
328.43
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
grade
pharmaceutical primary standard
agency
USP
API family
olanzapine
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
−70°C
SMILES string
[s]1c2c(cc1C)C(=Nc4c(cccc4)N2)N3CC[N](=O)(CC3)C
InChI
1S/C17H20N4OS/c1-12-11-13-16(20-7-9-21(2,22)10-8-20)18-14-5-3-4-6-15(14)19-17(13)23-12/h3-6,11,19H,7-10H2,1-2H3
InChI key
LJVNYCDXBXGQIK-UHFFFAOYSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Olanzapine Related Compound C USP reference standard intended for use in specified quality tests and assays.
Also used to prepare system suitability solutions for assay, and impurity analysis by liquid chromatography coupled to a UV detector according to the given below monographs of United States Pharmacopeia (USP):
Also used to prepare system suitability solutions for assay, and impurity analysis by liquid chromatography coupled to a UV detector according to the given below monographs of United States Pharmacopeia (USP):
- Olanzapine Orally Disintegrating Tablets
- Olanzapine Tablets
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
Still not finding the right product?
Explore all of our products under Olanzapine Related Compound C
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.