Skip to Content
Merck

Skip To

1466652

USP

N-Nitrosodiethylamine (NDEA)

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-Ethyl-N-nitrosoethanamine, NDEA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C4H10N2O
CAS Number:
Molecular Weight:
102.14
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

nifedipine

form

liquid

packaging

pkg of 1 mg

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical (small molecules)

format

neat

storage temp.

−20°C

SMILES string

N(N=O)(CC)CC

InChI

1S/C4H10N2O/c1-3-6(4-2)5-7/h3-4H2,1-2H3

InChI key

WBNQDOYYEUMPFS-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.

N-Nitrosodiethylamine is a carcinogenic compound that is strictly regulated within the biologics and pharmaceutical industries. Although not used as an ingredient, its presence is monitored in biologic products, as trace amounts can be toxic. NDEA is studied extensively for its potential health risks, and its detection is critical to ensuring the safety and regulatory compliance of biologic formulations.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

N-Nitrosodiethylamine (NDEA) USP reference standard is intended for use only as specificallyprescribed in the United States Pharmacopoeia.

It is also used to prepare standard, standard stock, nitrosamineRS stock, Nitrosamine standards stock solution mixture, and sensitivity stocksolutions to determine NDEA impurity in drug substances and drug products(valsartan, irbesartan, and losartan potassium etc.) by chromatography methodaccording to the general chapter <1469> of United States Pharmacopeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.
This product is part of USP Biologics Program.
This product is provided as delivered and specified by the USP Pharmacopoeia. For further information and support, including certificate/ product information sheets, please go to the website of the issuing Pharmacopoeia.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR2408Y000225840334
description

United States Pharmacopeia (USP) Reference Standard

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

European Pharmacopoeia (EP) Reference Standard

description

certified reference material, 5000 μg/mL in methanol

application(s)

USP Biologics
pharmaceutical (small molecules)

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

format

-

format

-

format

single component solution

form

liquid

form

liquid

form

-

form

-

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

certified reference material, TraceCERT®

storage temp.

−20°C

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

2-8°C

packaging

pkg of 1 mg

packaging

pkg of 1 g

packaging

-

packaging

ampule of 1 mL


Still not finding the right product?

Explore all of our products under N-Nitrosodiethylamine (NDEA)


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

49.5 °F

flash_point_c

9.7 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters