1460339
USP
Nepafenac Related Compound B
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
7-Benzoylindolin-2-one
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H11NO2
CAS Number:
Molecular Weight:
237.25
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
API family
nepafenac
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecules)
format
neat
storage temp.
2-8°C
InChI
1S/C15H11NO2/c17-13-9-11-7-4-8-12(14(11)16-13)15(18)10-5-2-1-3-6-10/h1-8H,9H2,(H,16,17)
InChI key
APGQYYFHBPQPTL-UHFFFAOYSA-N
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Application
Nepafenac Related Compound B USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service