Skip to Content
Merck
All Photos(1)

Key Documents

1435003

USP

Methylprednisolone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

6α-Methylprednisolone, 11β,17α,21-Trihydroxy-6α-methyl-1,4-pregnadiene-3,20-dione, 6α-Methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, Medrol, Medrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30O5
CAS Number:
Molecular Weight:
374.47
Beilstein:
2340300
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

methylprednisolone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12C[C@H](C)C3=CC(=O)C=C[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)CO

InChI

1S/C22H30O5/c1-12-8-14-15-5-7-22(27,18(26)11-23)21(15,3)10-17(25)19(14)20(2)6-4-13(24)9-16(12)20/h4,6,9,12,14-15,17,19,23,25,27H,5,7-8,10-11H2,1-3H3/t12-,14-,15-,17-,19+,20-,21-,22-/m0/s1

InChI key

VHRSUDSXCMQTMA-PJHHCJLFSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methylprednisolone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Methylprednisolone Sodium Succinate for Injection
  • Methylprednisolone Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Target Organs

Adrenal gland,Immune system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ornulf Paulsen et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 32(29), 3221-3228 (2014-07-09)
Corticosteroids are frequently used in cancer pain management despite limited evidence. This study compares the analgesic efficacy of corticosteroid therapy with placebo. Adult patients with cancer receiving opioids with average pain intensity ≥ 4 (numeric rating scale [NRS], 0 to
Emilio Portaccio et al.
Journal of neurology, neurosurgery, and psychiatry, 85(8), 845-850 (2014-01-10)
To assess relapses, disability progression and the role of disease modifying drugs (DMDs) in the year after delivery in women with multiple sclerosis (MS). We prospectively followed-up pregnancies occurring between 2002 and 2008 in women with MS, recruited from 21
Fungal infections associated with contaminated methylprednisolone injections.
Carol A Kauffman et al.
The New England journal of medicine, 368(26), 2495-2500 (2012-10-23)
Amy L Samuels et al.
British journal of haematology, 165(1), 57-66 (2014-01-25)
Drug-resistant forms of acute lymphoblastic leukaemia (ALL) are a leading cause of death from disease in children. Up to 25% of patients with T-cell ALL (T-ALL) develop resistance to chemotherapeutic agents, particularly to glucocorticoids (GCs), a class of drug to
M Toledano et al.
Neurology, 82(18), 1578-1586 (2014-04-08)
To evaluate a trial of immunotherapy as an aid to diagnosis in suspected autoimmune epilepsy. We reviewed the charts of 110 patients seen at our autoimmune neurology clinic with seizures as a chief complaint. Twenty-nine patients met the following inclusion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service