Skip to Content
Merck

Skip To

1426002

USP

Methyldopa

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Methyl-DOPA sesquihydrate, 3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine, MK-351, Methyl-L-DOPA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H13NO4 · 1.5H2O
CAS Number:
Molecular Weight:
238.24
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
2417244
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

methyldopa

manufacturer/tradename

USP

mp

≥300 °C

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[C@](N)(Cc1ccc(O)c(O)c1)C(O)=O

InChI

1S/C10H13NO4/c1-10(11,9(14)15)5-6-2-3-7(12)8(13)4-6/h2-4,12-13H,5,11H2,1H3,(H,14,15)/t10-/m0/s1

InChI key

CJCSPKMFHVPWAR-JTQLQIEISA-N

Gene Information

Application

Methyldopa USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0001083M1500000BP240
Methyldopa British Pharmacopoeia (BP) Reference Standard

BP240

Methyldopa

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

EDQM

manufacturer/tradename

BP

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

format

neat

format

neat

format

neat

Gene Information

human ... ADRA2A(150), ADRA2B(151), ADRA2C(152)

Gene Information

human ... ADRA2A(150), ADRA2B(151), ADRA2C(152)

Gene Information

human ... ADRA2A(150), ADRA2B(151), ADRA2C(152)

Gene Information

human ... ADRA2A(150), ADRA2B(151), ADRA2C(152)

mp

≥300 °C

mp

≥300 °C

mp

≥300 °C

mp

≥300 °C


Still not finding the right product?

Explore all of our products under Methyldopa


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters