Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C6H12N4
CAS Number:
Molecular Weight:
140.19
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
2018
grade
pharmaceutical primary standard
agency
USP
vapor pressure
<0.01 mmHg ( 20 °C)
API family
methenamine
autoignition temp.
770 °F
manufacturer/tradename
USP
mp
280 °C (subl.) (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
C1N2CN3CN1CN(C2)C3
InChI
1S/C6H12N4/c1-7-2-9-4-8(1)5-10(3-7)6-9/h1-6H2
InChI key
VKYKSIONXSXAKP-UHFFFAOYSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Methenamine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
Still not finding the right product?
Explore all of our products under Methenamine
signalword
Warning
hcodes
Hazard Classifications
Flam. Sol. 2 - Skin Sens. 1B
Storage Class
4.1B - Flammable solid hazardous materials
wgk
WGK 1
flash_point_f
482.0 °F - closed cup
flash_point_c
250 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

