Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
506719
grade
pharmaceutical primary standard
agency
USP
vapor density
3.72 (vs air)
vapor pressure
<1 mmHg ( 20 °C)
API family
amylmetacresol
autoignition temp.
1036 °F
expl. lim.
1.06-1.35 %, 150 °F
manufacturer/tradename
USP
refractive index
n
bp
203 °C (lit.)
mp
8-10 °C (lit.)
density
1.034 g/mL at 25 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
Cc1cccc(O)c1
InChI
1S/C7H8O/c1-6-3-2-4-7(8)5-6/h2-5,8H,1H3
InChI key
RLSSMJSEOOYNOY-UHFFFAOYSA-N
General description
Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
It is intended to be used as a USP reference standard in specified quality tests and assays.
Also used to prepare the standard solution in the assay and organic impurity analysis using gas chromatography-flame ionization detector (GC-FID) according to the United States Pharmacopeia (USP) monograph.
Also used to prepare the standard solution in the assay and organic impurity analysis using gas chromatography-flame ionization detector (GC-FID) according to the United States Pharmacopeia (USP) monograph.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk
WGK 2
flash_point_f
186.8 °F - closed cup
flash_point_c
86 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

