Skip to Content
Merck

Skip To

1379434

USP

Meloxicam Related Compound C

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Isopropyl 4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-dioxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H15NO5S
CAS Number:
Molecular Weight:
297.33
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

meloxicam

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/C13H15NO5S/c1-8(2)19-13(16)11-12(15)9-6-4-5-7-10(9)20(17,18)14(11)3/h4-8,15H,1-3H3

InChI key

ZHWSBDIYCSOTBY-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Meloxicam Related Compound C USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Meloxicam

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR25021379445PHR3395
grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

USP

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

format

neat

format

-

format

neat

format

-

API family

meloxicam

API family

meloxicam

API family

meloxicam

API family

-


Still not finding the right product?

Explore all of our products under Meloxicam Related Compound C


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters