Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
grade
pharmaceutical primary standard
agency
USP
API family
magnesium chloride
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
O.O.O.O.O.O.Cl[Mg]Cl
InChI
1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2
InChI key
DHRRIBDTHFBPNG-UHFFFAOYSA-L
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Magnesium chloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename - | manufacturer/tradename - |
| grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade - | grade - |
| format neat | format - | format - | format - |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical small molecule | application(s) pharmaceutical (small molecule) | application(s) liquid formulation |
| API family magnesium chloride | API family magnesium chloride | API family - | API family - |
Still not finding the right product?
Explore all of our products under Magnesium chloride
Storage Class
13 - Non Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
