Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C15H8Cl2N2O
CAS Number:
Molecular Weight:
303.14
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
grade
pharmaceutical primary standard
agency
USP
API family
lorazepam
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
Clc1ccc2nc(C=O)nc(-c3ccccc3Cl)c2c1
InChI
1S/C15H8Cl2N2O/c16-9-5-6-13-11(7-9)15(19-14(8-20)18-13)10-3-1-2-4-12(10)17/h1-8H
InChI key
VUUITUUENPOMIU-UHFFFAOYSA-N
Application
Lorazepam Related Compound C USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
- Lorazepam Oral Concentrate
- Lorazepam Injection
- Lorazepam
- Lorazepam Tablets
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
Still not finding the right product?
Explore all of our products under Lorazepam Related Compound C
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Chronic 4 - Eye Irrit. 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
