Skip to Content
Merck

1370338

USP

Lorazepam Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Amino-2′,5-dichlorobenzophenone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
H2NC6H3(Cl)COC6H4Cl
CAS Number:
Molecular Weight:
266.12
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

lorazepam

manufacturer/tradename

USP

mp

87-89 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Nc1ccc(Cl)cc1C(=O)c2ccccc2Cl

InChI

1S/C13H9Cl2NO/c14-8-5-6-12(16)10(7-8)13(17)9-3-1-2-4-11(9)15/h1-7H,16H2

InChI key

KWZYIAJRFJVQDO-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.

Application

Lorazepam Related Compound B USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Lorazepam Oral Concentrate
  • Lorazepam Injection
  • Lorazepam
  • Lorazepam Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library