Skip to Content
Merck
All Photos(1)

Documents

1366080

USP

Lidocaine Related Compound H

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-(2,6-Dimethylphenyl)chloroacetamide, 1-Chloroacetylamino-2,6-dimethylbenzene, 2-Chloro-2′,6′-acetoxylidide, 2-Chloro-2′,6′-dimethylacetanilide, 2-Chloro-N-(2,6-dimethylphenyl)acetamide, N-(2,6-Dimethylphenyl)-2-chloroacetamide, N-(2,6-Xylyl)chloroacetamide, N-Chloroacetyl-2,6-dimethylaniline, Chloroacetamido-2,6-xylidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CONHC6H3(CH3)2
CAS Number:
Molecular Weight:
197.66
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lidocaine

manufacturer/tradename

USP

mp

150-151 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cc1cccc(C)c1NC(=O)CCl

InChI

1S/C10H12ClNO/c1-7-4-3-5-8(2)10(7)12-9(13)6-11/h3-5H,6H2,1-2H3,(H,12,13)

InChI key

FPQQSNUTBWFFLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lidocaine Related Compound H USP reference standard for specifies quality tests and assay use.

Also used to prepare standard solutions for impurity analysis according to the given below monographs of United States Pharmacopeia (USP):
  • Lidocaine
  • Lidocaine Hydrochloride Topical Solution
  • Lidocaine Hydrochloride Oral Topical Solution
  • Lidocaine Hydrochloride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Lidocaine Hydrochloride
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(3), 2623-2623 (2020)
Lidocaine
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(3), 2620-2620 (2020)
Lidocaine Hydrochloride Oral Topical Solution
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(4), 2626-2626 (2020)
Lidocaine Hydrochloride Topical Solution
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(4), 2627-2627 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service