Skip to Content
Merck
All Photos(1)

Key Documents

1356632

USP

Ketoprofen

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-(3-Benzoylphenyl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O3
CAS Number:
Molecular Weight:
254.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ketoprofen

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C(O)=O)c1cccc(c1)C(=O)c2ccccc2

InChI

1S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11H,1H3,(H,18,19)

InChI key

DKYWVDODHFEZIM-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ketoprofen USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Ketoprofen Capsules
  • Ketoprofen Extended-Release Capsules

Biochem/physiol Actions

Non-steroidal anti-inflammatory compound that is selective for COX-1.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jodie Barden et al.
The Cochrane database of systematic reviews, (4)(4), CD007355-CD007355 (2009-10-13)
Ketoprofen is a non-selective non-steroidal anti-inflammatory drug (NSAID) used to treat acute and chronic painful conditions. Dexketoprofen is the (S)-enantiomer, which is believed to confer analgesia. Theoretically dexketoprofen is expected to provide equivalent analgesia to ketoprofen at half the dose
F Jamali et al.
Clinical pharmacokinetics, 19(3), 197-217 (1990-09-01)
Ketoprofen, a potent nonsteroidal anti-inflammatory drug (NSAID) of the 2-arylpropionic acid class, has been used clinically for over 15 years in Europe, and has recently been introduced in the United States. Although it possesses a chiral centre, with only the
Fabíola Garavello Prezotti et al.
Carbohydrate polymers, 113, 286-295 (2014-09-27)
Gellan gum/pectin beads were prepared by ionotropic gelation, using Al(3+) as crosslinker. High yield (92.76%) and entrapment efficiency (52.22-88.78%) were reached. Beads exhibited high circularity (0.730-0.849) and size between 728.95 and 924.56 μm. Particle size and circularity was increased by
H Bagheri et al.
Drug safety, 22(5), 339-349 (2000-06-01)
The topical use of nonsteroidal anti-inflammatory drugs (NSAIDs), widely used for moderate acute and chronic painful conditions, is one of several strategies used to improve the tolerability profile of NSAIDs, particularly with regard to gastric and renal adverse effects. However
Akihiko Ouchi et al.
Chirality, 14(1), 1-11 (2001-12-19)
The reactivity of organic molecules can be modified upon complexation with proteins: these changes can be different and more significant when the substrate is in an electronically excited state. Here we review UV, CD, and fluorescence spectroscopy studies on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service