Skip to Content
Merck

1302214

USP

Guggulsterone Z

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(Z)-Guggulsterone, (17Z)-Pregna-4,17(20)-diene-3,16-dione, 4,17(20)-trans-Pregnadiene-3,16-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

guggul

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)\C(=C\C)C(=O)C[C@@]24[H]

InChI

1S/C21H28O2/c1-4-16-19(23)12-18-15-6-5-13-11-14(22)7-9-20(13,2)17(15)8-10-21(16,18)3/h4,11,15,17-18H,5-10,12H2,1-3H3/b16-4+/t15-,17+,18+,20+,21-/m1/s1

InChI key

WDXRGPWQVHZTQJ-OSJVMJFVSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Guggulsterone Z USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Guggul
  • Guggul Tablets
  • Native Guggul Extract
  • Purified Guggul Extract

Biochem/physiol Actions

(Z)-Guggulsterone is a natural product that lowers cholesterol due to its function as an antagonist ligand for the bile acid receptor. (Z)-Guggulsterone is a nuclear hormone receptor that regulates the transcription of several genes involved in cholesterol metabolism and plays a role in cholesterol level regulation. (Z)-Guggulsterone is also a selective farnesoid X receptor (FXR) modulator.
(Z)-Guggulsterone is an antagonist ligand for bile acid receptor.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Guggulsterone Z


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library