Skip to Content
Merck
All Photos(1)

Key Documents

1287700

USP

Galactose

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

α-D-Galactose, α-D-Galactopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

galactose

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O1[C@@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O)O)O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3+,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-PHYPRBDBSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Galactose contains d-galactopyranose (C6H12O6), which is obtained either as one of the products of the metabolism of lactose, a naturally occurring sugar in dairy products, by the digestive enzyme lactase, or from arabinogalactans isolated from a plant-derived source.

Application

Galactose USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Guar Gum
  • Lactulose Concentrate
  • Lactulose Solution
  • Ganoderma Lucidum Fruiting Body Powder
  • Ganoderma Lucidum Fruiting Body

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lactulose Concentrate
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(5), 2521-2521 (2016)
Ganoderma Lucidum Fruiting Body Powder
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(5), 5008-5008 (2015)
Guar Gum
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(2), 5813-5813 (2018)
Galactose
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(5) (2020)
Michelle P Christie et al.
PloS one, 9(4), e95024-e95024 (2014-04-17)
Glycosylation of biopharmaceuticals can mediate cell specific delivery by targeting carbohydrate receptors. Additionally, glycosylation can improve the physico-chemical (drug-like) properties of peptide based drug candidates. The main purpose of this study was to examine if glycosylation of the peptide enkephalin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service