Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
furazolidone
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
[O-][N+](=O)c1ccc(\C=N\N2CCOC2=O)o1
InChI
1S/C8H7N3O5/c12-8-10(3-4-15-8)9-5-6-1-2-7(16-6)11(13)14/h1-2,5H,3-4H2/b9-5+
InChI key
PLHJDBGFXBMTGZ-WEVVVXLNSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename - | manufacturer/tradename USP |
| grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade - | grade pharmaceutical primary standard |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical small molecule | application(s) - | application(s) pharmaceutical (small molecule) |
| format neat | format - | format - | format neat |
| API family furazolidone | API family furazolidone | API family - | API family nitrofurazone |
Still not finding the right product?
Explore all of our products under Furazolidone
signalword
Warning
hcodes
Hazard Classifications
Repr. 2
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


