Skip to Content
Merck
All Photos(1)

Key Documents

1286504

USP

Fructose

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

D-(−)-Fructose, D-Levulose, Fruit sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1239004
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fructose

manufacturer/tradename

USP

mp

119-122 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m1/s1

InChI key

BJHIKXHVCXFQLS-UYFOZJQFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Fructose, dried under vacuum at 70° for 4 h.

Application

Fructose USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Fructose Injection
  • High Fructose Corn Syrup
  • Fructose and Sodium Chloride Injection
  • Lactulose Concentrate
  • D-Chiro-Inositol
  • Compressible Sugar

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fructose Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 2049-2049 (2018)
Xylose
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 34(4), 4662-4662 (2020)
Fructose
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(5) (2022)
Inulin
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 29(6), 2360-2360 (2018)
High Fructose Corn Syrup
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(1), 5727-5727 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service