Skip to Content
Merck

1286107

USP

Formoterol fumarate

United States Pharmacopeia (USP) Reference Standard, monograph mol wt. 840.91 ((C19H24N2O4)2 · C4H4O4 ·2H2O)

Synonym(s):

Formoterol fumarate dihydrate, ((±)-2′-Hydroxy-5′-[(R*)-1-hydroxy-2-[[(R*)-p-methoxy-a-methylphenethyl]amino]ethyl]formanilide fumarate (2:1) (salt) dihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
(C19H24N2O4)2 · C4H4O4 · 2H2O
CAS Number:
Molecular Weight:
840.91
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

formoterol 

mol wt

monograph mol wt. 840.91

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N(C(Cc4ccc(cc4)OC)C)CC(O)c3cc(c(cc3)O)NC=O.N(C(Cc2ccc(cc2)OC)C)CC(O)c1cc(c(cc1)O)NC=O.OC(=O)\C=C\C(=O)O.O.O

InChI

1S/2C19H24N2O4.C4H4O4.2H2O/c2*1-13(9-14-3-6-16(25-2)7-4-14)20-11-19(24)15-5-8-18(23)17(10-15)21-12-22;5-3(6)1-2-4(7)8;;/h2*3-8,10,12-13,19-20,23-24H,9,11H2,1-2H3,(H,21,22);1-2H,(H,5,6)(H,7,8);2*1H2/b;;2-1+;;

InChI key

RATSWNOMCHFQGJ-XODSYJLDSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Application

This USP reference standard is intended for use in specified quality tests and assays.
Also used to prepare standard and system suitability solutions for the assay and impurity analysis according to the United States Pharmacopeia (USP) monograph.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Formoterol fumarate


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Repr. 2 - STOT RE 2 - STOT SE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library