Skip to Content
Merck

1247003

USP

Erythromycin lactobionate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Erythromycin 4-O-β-D-galactopyranosyl-D-gluconate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C37H67NO13 · C12H22O12
CAS Number:
Molecular Weight:
1092.22
MDL number:
UNSPSC Code:
41116107
EC Number:
223-348-7
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

erythromycin

form

powder

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

SMILES string

N([C@@H]2[C@H]([C@@H](O[C@@H](C2)C)O[C@@H]3[C@H]([C@@H]([C@H](C(=O)O[C@@H]([C@]([C@@H]([C@H](C(=O)[C@@H](C[C@]3(O)C)C)C)O)(O)C)CC)C)O[C@@H]4O[C@H]([C@@H]([C@@](C4)(OC)C)O)C)C)O)(C)C.O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O)O)O[C@@H]([C@H](O)[C@@H](O)C(=O)O

InChI

1S/C37H67NO13.C12H22O12/c1-14-25-37(10,45)30(41)20(4)27(39)18(2)16-35(8,44)32(51-34-28(40)24(38(11)12)15-19(3)47-34)21(5)29(22(6)33(43)49-25)50-26-17-36(9,46-13)31(42)23(7)48-26;13-1-3(15)10(7(18)8(19)11(21)22)24-12-9(20)6(17)5(16)4(2-14)23-12/h18-26,28-32,34,40-42,44-45H,14-17H2,1-13H3;3-10,12-20H,1-2H2,(H,21,22)/t18-,19-,20+,21+,22-,23+,24+,25-,26+,28-,29+,30-,31+,32-,34+,35-,36-,37-;3-,4-,5+,6+,7-,8-,9-,10-,12+/m11/s1

InChI key

NNRXCKZMQLFUPL-WBMZRJHASA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.

Erythromycin lactobionate is a highly water-soluble form of erythromycin used primarily in intravenous preparations. It retains the macrolide′s mechanism of action, targeting the 50S ribosomal subunit. The USP reference standard supports analytical testing for intravenous dosage forms.

The USP biologics antibiotics category includes a wide range of antimicrobial agents that are essential in treating bacterial infections. These antibiotics are derived from various natural sources or synthesized to combat specific pathogens effectively. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of antibiotic therapeutics throughout their lifecycle.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

Erythromycin lactobionate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Erythromycin Lactobionate for Injection
  • Sterile Erythromycin Lactobionate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
This product is part of the USP Biologics program.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library