Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
ergosterol
manufacturer/tradename
USP
mp
156-158 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
[H][C@@]1(CC[C@@]2([H])C3=CC=C4C[C@@H](O)CC[C@]4(C)[C@@]3([H])CC[C@]12C)[C@H](C)\C=C\[C@H](C)C(C)C
InChI
1S/C28H44O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h7-10,18-20,22,24-26,29H,11-17H2,1-6H3/b8-7+/t19-,20+,22-,24+,25-,26-,27-,28+/m0/s1
InChI key
DNVPQKQSNYMLRS-APGDWVJJSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Also, for use with USP monographs such as:
- Ganoderma Lucidum Fruiting Body Powder
- Ganoderma Lucidum Fruiting Body
- Ergocalciferol
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade - |
| manufacturer/tradename USP | manufacturer/tradename EDQM | manufacturer/tradename - | manufacturer/tradename - |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) - |
| format neat | format neat | format neat | format - |
| mp 156-158 °C (lit.) | mp 156-158 °C (lit.) | mp 156-158 °C (lit.) | mp 156-158 °C (lit.) |
| API family ergosterol | API family ergosterol | API family ergosterol | API family - |
Still not finding the right product?
Explore all of our products under Ergosterol
hcodes
Hazard Classifications
Aquatic Chronic 4
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
