Skip to Content
Merck

1240004

USP

Ergonovine maleate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Ergonovine maleate salt, Ergometrine maleate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H23N3O2 · C4H4O4
CAS Number:
Molecular Weight:
441.48
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

ergometrine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)\C=C/C(O)=O.C[C@@H](CO)NC(=O)[C@H]1CN(C)[C@@H]2Cc3c[nH]c4cccc(C2=C1)c34

InChI

1S/C19H23N3O2.C4H4O4/c1-11(10-23)21-19(24)13-6-15-14-4-3-5-16-18(14)12(8-20-16)7-17(15)22(2)9-13;5-3(6)1-2-4(7)8/h3-6,8,11,13,17,20,23H,7,9-10H2,1-2H3,(H,21,24);1-2H,(H,5,6)(H,7,8)/b;2-1-/t11-,13+,17+;/m0./s1

InChI key

YREISLCRUMOYAY-IIPCNOPRSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ergonovine maleate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Ergonovine Maleate Injection

Biochem/physiol Actions

Oxytocic, potent antagonist at smooth muscle serotonergic receptors.
Oxytocic, stimulates contraction of uterine muscle; partial agonist at serotonergic and dopaminergic receptors in CNS; selective, potent antagonist at smooth muscle serotonergic receptors.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1240004-150MG04061833844700
1240004-100MG04061833830383