Skip to Content
Merck

1239005

USP

Ergocalciferol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Calciferol, Ercalciol, Ergosterol irradiated, Irradiated ergosterol, Vitamin D2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H44O
CAS Number:
Molecular Weight:
396.65
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1916682
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

ergocalciferol

manufacturer/tradename

USP

mp

114-118 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CC(C)[C@@H](C)/C=C/[C@@H](C)[C@@]1([H])CC[C@@]([C@]1(C)CCC/2)([H])C2=C\C=C(C[C@@H](O)CC3)/C3=C

InChI

1S/C28H44O/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(29)14-11-21(24)4/h9-10,12-13,19-20,22,25-27,29H,4,7-8,11,14-18H2,1-3,5-6H3/b10-9+,23-12+,24-13-/t20-,22+,25-,26+,27-,28+/m0/s1

InChI key

MECHNRXZTMCUDQ-RKHKHRCZSA-N

Gene Information

human ... VDR(7421)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Ergocalciferol


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Tom D Thacher et al.
Archives of disease in childhood, 99(9), 807-811 (2014-04-22)
To determine whether children with calcium-deficiency rickets have a better response to treatment with vitamin D and calcium than with calcium alone. Randomised controlled trial. Jos University Teaching Hospital, Jos, Nigeria. Nigerian children with active rickets treated with calcium carbonate
James E Heubi et al.
Hepatology (Baltimore, Md.), 61(1), 268-274 (2014-08-29)
Bile acid amidation defects were predicted to present with fat/fat soluble vitamin malabsorption with minimal cholestasis. We identified and treated five patients (one male, four females) from four families with defective bile acid amidation due to a genetically confirmed deficiency
Daniela Max et al.
Molecular nutrition & food research, 58(2), 343-352 (2013-08-22)
Epidemiologic data reveal associations between low serum concentrations of 25-hydroxyvitamin D (25(OH)D) and higher risk of falls and muscle weakness. Fetal stage is critical for the development of skeletal muscle, but little information is available on the impact of maternal



Global Trade Item Number

SKUGTIN
1239005-5X30MG04061838722645