Skip to Content
Merck

1237382

USP

Epirubicin hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4′-Epidoxorubicin hydrochloride, Epidoxorubicin hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
C27H29NO11 · HCl
CAS Number:
Molecular Weight:
579.98
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Epirubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard

InChI

1S/C27H29NO11.ClH/c1-10-22(31)13(28)6-17(38-10)39-15-8-27(36,16(30)9-29)7-12-19(15)26(35)21-20(24(12)33)23(32)11-4-3-5-14(37-2)18(11)25(21)34;/h3-5,10,13,15,17,22,29,31,33,35-36H,6-9,28H2,1-2H3;1H/t10-,13-,15-,17-,22-,27-;/m0./s1

SMILES string

Cl.COc1cccc2C(=O)c3c(O)c4C[C@](O)(C[C@H](O[C@H]5C[C@H](N)[C@@H](O)[C@H](C)O5)c4c(O)c3C(=O)c12)C(=O)CO

InChI key

MWWSFMDVAYGXBV-FGBSZODSSA-N

grade

pharmaceutical primary standard

API family

epirubicin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
E0550000BP992E9406
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

BP

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

-

format

neat

format

neat

format

neat

format

-

API family

epirubicin

API family

epirubicin

API family

epirubicin

API family

-

storage temp.

−20°C

storage temp.

-

storage temp.

−20°C

storage temp.

−20°C

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Application

Epirubicin hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Doxorubicin Hydrochloride
  • Doxorubicin Hydrochloride for Injection
  • Doxorubicin Hydrochloride Injection
  • Epirubicin Hydrochloride
  • Epirubicin Hydrochloride Injection

Biochem/physiol Actions

Cell-permeable anthracycline antitumor antibiotic. Antineoplastic. A stereoisomer of doxorubicin that exhibits reduced cardiotoxicity. Its antitumor actions are mediated by targeting topoisomerase II.

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Other Notes

Sales restrictions may apply.

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Muta. 1B - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service