Skip to Content
Merck
All Photos(1)

Documents

1235900

USP

Enrofloxacin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Baytril

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22FN3O3
CAS Number:
Molecular Weight:
359.39
Beilstein:
5307824
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

enrofloxacin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCN1CCN(CC1)c2cc3N(C=C(C(O)=O)C(=O)c3cc2F)C4CC4

InChI

1S/C19H22FN3O3/c1-2-21-5-7-22(8-6-21)17-10-16-13(9-15(17)20)18(24)14(19(25)26)11-23(16)12-3-4-12/h9-12H,2-8H2,1H3,(H,25,26)

InChI key

SPFYMRJSYKOXGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Enrofloxacin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monograph such as Enrofloxacin Compounded Oral Suspension, Veterinary

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jochen Springer et al.
Stroke, 45(12), 3675-3683 (2014-10-30)
Muscle wasting is a common complication accompanying stroke. Although it is known to impair poststroke recovery, the mechanisms of subacute catabolism after stroke have not been investigated in detail. The aim of this study is to investigate mechanisms of local
Brian T David et al.
Journal of neurotrauma, 31(21), 1800-1806 (2014-06-18)
Spinal cord injury (SCI) affects motor, sensory, and autonomic functions. As current therapies do not adequately alleviate functional deficits, the development of new and more effective approaches is of critical importance. Our earlier investigations indicated that intrathecal administration of a
Taylor B Engle et al.
BMC veterinary research, 10, 286-286 (2014-12-05)
Porcine circovirus 2 is the primary agent responsible for inducing a group of associated diseases known as Porcine Circovirus Associated Diseases (PCVAD), which can have detrimental effects on production efficiency as well as causing significant mortality. The objective of this
Susanne Hoffmann et al.
Journal of neurophysiology, 113(4), 1135-1145 (2014-11-21)
Locomotion and foraging on the wing require precise navigation in more than just the horizontal plane. Navigation in three dimensions and, specifically, precise adjustment of flight height are essential for flying animals. Echolocating bats drink from water surfaces in flight
S C Coelho et al.
European journal of pain (London, England), 18(7), 957-967 (2014-01-09)
This study evaluated the involvement of tumour necrosis factor α (TNF-α) in orofacial thermal and mechanical hyperalgesia induced by an inflammatory stimulus or by chronic constriction of the infraorbital nerve (CION) using etanercept (Eta), a TNF-receptor fusion protein that inhibits

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service