Skip to Content

1187080

USP

Dibutyl phthalate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

n-Butyl phthalate, DBP, Phthalic acid dibutyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H4-1,2-[CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
278.34
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1914064
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor density

9.6 (vs air)

vapor pressure

1 mmHg ( 147 °C)

API family

dibutyl phthalate

form

liquid

autoignition temp.

756 °F

expl. lim.

0.47 %, 236 °F

manufacturer/tradename

USP

refractive index

n20/D 1.492 (lit.)

bp

340 °C (lit.)

mp

−35 °C (lit.)

density

1.043 g/mL at 25 °C (lit.)

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

SMILES string

CCCCOC(=O)c1ccccc1C(=O)OCCCC

InChI

1S/C16H22O4/c1-3-5-11-19-15(17)13-9-7-8-10-14(13)16(18)20-12-6-4-2/h7-10H,3-6,11-12H2,1-2H3

InChI key

DOIRQSBPFJWKBE-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.

Dibutyl phthalate (DBP) is a plasticizer commonly used in the formulation of biologics to improve the flexibility and stability of polymeric materials. It plays an essential role in the production of drug delivery devices and packaging for biologics. By enhancing the physical properties of polymers, DBP contributes to the reliability and performance of biologic products, particularly those in extended-release or controlled-release systems.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

This compound is listed in the SVHC (Substances of very high concern) candidate list of ECHA (European Chemicals Agency)
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Plasticizer

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
This compound is listed in the SVHC (Substances of very high concern) candidate list of ECHA (European Chemicals Agency)This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
This product is part of USP Biologics Program.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pha


Still not finding the right product?

Explore all of our products under Dibutyl phthalate


pictograms

Health hazardEnvironment

signalword

Danger

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

367.7 °F - open cup

flash_point_c

186.5 °C - open cup

Hazard Classifications

Aquatic Acute 1 - ED ENV 1 - ED HH 1 - Repr. 1B



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library