Skip to Content
Merck
All Photos(1)

Key Documents

1173508

USP

Desoximetasone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

9-Fluoro-11β,21-dihydroxy-16α-methylpregna-1,4-diene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29FO4
CAS Number:
Molecular Weight:
376.46
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

biological source

synthetic

Agency

USP

vapor pressure

<0.0000001 kPa ( 25 °C)

API family

desoximetasone

form

powder

packaging

pkg of 200 mg

manufacturer/tradename

USP

storage condition

protect from light

color

white

solubility

acetone: freely soluble
alcohol: freely soluble
benzene: slightly soluble
chloroform: freely soluble
ether: slightly soluble
water: insoluble

application(s)

pharmaceutical (small molecule)

SMILES string

[H][C@@]12C[C@@H](C)[C@H](C(=O)CO)[C@@]1(C)C[C@H](O)[C@@]3(F)[C@@]2([H])CCC4=CC(=O)C=C[C@]34C

InChI

1S/C22H29FO4/c1-12-8-16-15-5-4-13-9-14(25)6-7-21(13,3)22(15,23)18(27)10-20(16,2)19(12)17(26)11-24/h6-7,9,12,15-16,18-19,24,27H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,18+,19-,20+,21+,22+/m1/s1

InChI key

VWVSBHGCDBMOOT-IIEHVVJPSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Desoximetasone is a topical corticosteroid known to modify the functions of leukocytes that are mainly responsible for skin infections, inflammation and allergies.

Application

Desoximetasone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Desoximetasone Cream
  • Desoximetasone Gel
  • Desoximetasone Ointment
  • Dexamethasone

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and Validation of RP-HPLC Method for Desoximetasone in Bulk and Cream Formulation
Gorle A, et al.
Journal of Drug Delivery and Therapeutics, 9(3), 154-159 (2019)
Topical corticosteroids: mechanisms of action.
Kragballe, K
Acta Dermato-Venereologica. Supplementum, 151(3), 7-7 (1989)
S Vladimirov et al.
Journal of pharmaceutical and biomedical analysis, 14(8-10), 947-950 (1996-06-01)
The proposed method is based on coloured hydrazone formation with 1,4-dihydrazinophthalazine as a reagent. Heating at 85 degrees C for 2 h was found necessary to ensure optimal hydrazone formation in the presence of hydrochloric acid. The yellow hydrazone product
Desoximetasone USP Monograph
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(2), 1272-1272 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service