Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
form
neat
packaging
pkg of 20 mg
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
pharmaceutical
pharmaceutical
format
neat
General description
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description CRS, European Pharmacopoeia (EP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description European Pharmacopoeia (EP) Reference Standard |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| manufacturer/tradename USP | manufacturer/tradename EDQM | manufacturer/tradename USP | manufacturer/tradename EDQM |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| form neat | form - | form - | form - |
| format neat | format - | format neat | format neat |
| agency USP | agency BP | agency USP | agency - |
Still not finding the right product?
Explore all of our products under Croscarmellose sodium
Storage Class
11 - Combustible Solids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
