Skip to Content
Merck
All Photos(1)

Key Documents

1098130

USP

Ceftazidime pentahydrate

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22N6O7S2 · 5H2O
CAS Number:
Molecular Weight:
636.65
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ceftazidime

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[s]1c(nc(c1)\C(=N\OC(C)(C)C(=O)[O-])\C(=O)N[C@H]2[C@H]3SCC(=C(N3C2=O)C(=O)[O-])C[n+]4ccccc4)N.O.O.O.O.O.[H+]

InChI

1S/C22H22N6O7S2.5H2O/c1-22(2,20(33)34)35-26-13(12-10-37-21(23)24-12)16(29)25-14-17(30)28-15(19(31)32)11(9-36-18(14)28)8-27-6-4-3-5-7-27;;;;;/h3-7,10,14,18H,8-9H2,1-2H3,(H4-,23,24,25,29,31,32,33,34);5*1H2/b26-13-;;;;;/t14-,18-;;;;;/m1...../s1

InChI key

NMVPEQXCMGEDNH-TZVUEUGBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ceftazidime pentahydrate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Ceftazidime
  • Ceftazidime Injection
  • Ceftazidime for Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceftazidime
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 45(3), 872-872 (2018)
M A Sirgo et al.
DICP : the annals of pharmacotherapy, 25(3), 284-288 (1991-03-01)
The disposition of drugs in the elderly is particularly relevant with antiinfectives, because this population has an increased risk of infections. Renal function deteriorates with age, yet dosage guidelines for antibiotics that allow for this reduction remain to be established.
George G Zhanel et al.
Drugs, 73(2), 159-177 (2013-02-02)
Avibactam (formerly NXL104, AVE1330A) is a synthetic non-β-lactam, β-lactamase inhibitor that inhibits the activities of Ambler class A and C β-lactamases and some Ambler class D enzymes. This review summarizes the existing data published for ceftazidime-avibactam, including relevant chemistry, mechanisms
A Walkty et al.
Antimicrobial agents and chemotherapy, 57(11), 5707-5709 (2013-08-14)
The in vitro activity of ceftolozane in combination with tazobactam (fixed concentration of 4 μg/ml) was evaluated against 2,435 Pseudomonas aeruginosa clinical isolates obtained from across Canada using Clinical and Laboratory Standards Institute broth microdilution methods. The MIC50 and MIC90
Gavin C K W Koh et al.
PLoS neglected tropical diseases, 7(10), e2500-e2500 (2013-10-23)
Burkholderia pseudomallei infection (melioidosis) is an important cause of community-acquired Gram-negative sepsis in Northeast Thailand, where it is associated with a ~40% mortality rate despite antimicrobial chemotherapy. We showed in a previous cohort study that patients taking glyburide ( =

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service