Skip to Content
Merck
All Photos(1)

Documents

1098027

USP

Cefpodoxime proxetil

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N5O9S2
CAS Number:
Molecular Weight:
557.60
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cefpodoxime

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C21H27N5O9S2/c1-9(2)33-21(30)35-10(3)34-19(29)15-11(6-31-4)7-36-18-14(17(28)26(15)18)24-16(27)13(25-32-5)12-8-37-20(22)23-12/h8-10,14,18H,6-7H2,1-5H3,(H2,22,23)(H,24,27)/b25-13-/t10?,14-,18-/m1/s1

InChI key

LTINZAODLRIQIX-FBXRGJNPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cefpodoxime proxetil USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Cefpodoxime Proxetil for Oral Suspension
  • Cefpodoxime Proxetil Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amrita Bajaj et al.
Drug development and industrial pharmacy, 39(5), 635-645 (2012-05-09)
Lipid based drug delivery systems have gained prominence in last decade for drugs with dissolution rate limited oral bioavailability. To improve the solubility, permeability and oral bioavailability of cefpodoxime proxetil, β-lactam antibiotic. It is BCS Class IV drug having solubility
S Bhargava et al.
Current drug delivery, 5(1), 1-6 (2008-01-29)
Cefpodoxime proxetil (CPDX-PR) is an oral cephalosporin antibiotic with poor aqueous solubility and bioavailability. Effect of beta-cyclodextrin on aqueous solubility and dissolution rate of cefpodoxime proxetil was evaluated by the formation of solid inclusion complexes in 1:2 molar ratio of
Md Salim Shakur et al.
Indian pediatrics, 44(11), 838-841 (2007-12-07)
In order to evaluate clinical and bacteriological efficacy of Cefpodoxime Proxetil (CP) in typhoid fever in comparison to cefixime (CF), we assessed 140 children with suspected typhoid fever. Fulfilling inclusion criteria finally 40 culture confirmed typhoid fever were allocated in
Santosh V Gandhi et al.
Hindustan antibiotics bulletin, 51(1-4), 24-28 (2009-01-01)
Two accurate, precise, sensitive and economical procedures for simultaneous estimation of Cefpodoxime proxetil and Potassium clavulanate in tablet dosage form have been developed. The methods employed were absorbance correction method (I) and first order derivative spectroscopic method (II). The first
Abhijit A Date et al.
International journal of pharmaceutics, 329(1-2), 166-172 (2006-10-03)
Self-nanoemulsifying drug delivery systems (SNEDDS) were developed with the objective to overcome problems associated with the delivery of cefpodoxime proxetil (CFP), a poorly bioavailable high dose antibiotic having pH dependant solubility. Solubility of CFP in oily phases and surfactants was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service