Skip to Content
Merck
All Photos(1)

Documents

1091200

USP

Captopril

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-[(S)-3-Mercapto-2-methylpropionyl]-L-proline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15NO3S
CAS Number:
Molecular Weight:
217.29
Beilstein:
477887
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

captopril

manufacturer/tradename

USP

mp

104-108 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[C@H](CS)C(=O)N1CCC[C@H]1C(O)=O

InChI

1S/C9H15NO3S/c1-6(5-14)8(11)10-4-2-3-7(10)9(12)13/h6-7,14H,2-5H2,1H3,(H,12,13)/t6-,7+/m1/s1

InChI key

FAKRSMQSSFJEIM-RQJHMYQMSA-N

Gene Information

human ... ACE(1636)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Captopril USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Captopril and Hydrochlorothiazide Tablets
  • Captopril Compounded Oral Solution
  • Captopril Compounded Oral Suspension
  • Captopril Tablets

Biochem/physiol Actions

Angiotensin converting enzyme inhibitor. Inhibits the formation of angiotensin II, a bioactive peptide that stimulates angiogenesis and increases microvessel density.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R I Ogilvie et al.
The Canadian journal of cardiology, 14(8), 1025-1033 (1998-09-17)
Twenty-four splenectomized dogs were subjected to rapid right ventricular pacing (RRVP) at 250 beats/min for five weeks. During the final three weeks, four groups six dogs were untreated or treated with captopril alone, with the angiotensin II type 1 (AT1)
Antonio C M Camargo et al.
Toxicon : official journal of the International Society on Toxinology, 59(4), 516-523 (2011-08-13)
The identification of novel endogenous and exogenous molecules acting in the complex mechanism of regulating the vascular tonus has always been of great interest. The discovery of bradykinin (1949) and the bradykinin-potentiating peptides (1965) had a pivotal influence in the
Javed Mahmood et al.
International journal of radiation oncology, biology, physics, 89(4), 722-728 (2014-05-29)
To investigate the outcome of suppression of the renin angiotensin system using captopril combined with an antioxidant (Eukarion [EUK]-207) for mitigation of radiation-induced lung damage in rats. The thoracic cavity of female Sprague-Dawley rats was irradiated with a single dose
Kyung Hee Chang et al.
Nature communications, 6, 5914-5914 (2015-01-13)
Patients with organ failure of vascular origin have increased circulating haematopoietic stem cells and progenitors (HSC/P). Plasma levels of angiotensin II (Ang-II), are commonly increased in vasculopathies. Hyperangiotensinemia results in activation of a very distinct Ang-II receptor set, Rho family
Johannes J Kovarik et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 30(1), 115-123 (2014-08-12)
Blockade of the renin-angiotensin system (RAS) exerts beneficial effects in patients with mild-to-moderate chronic kidney disease, yet evidence suggesting a similar benefit in haemodialysis (HD) patients is not available. Furthermore, knowledge of the effects of RAS blockade on systemic RAS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service