Skip to Content
Merck
All Photos(1)

Key Documents

1044516

USP

Atorvastatin Calcium

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Atorvastatin hemicalcium salt sesquihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H68CaF2N4O10 · 3H2O
CAS Number:
Molecular Weight:
1209.39
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

atorvastatin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Ca+2].Fc5ccc(cc5)c6[n](c(c(c6c8ccccc8)C(=O)Nc7ccccc7)C(C)C)CC[C@@H](O)C[C@@H](O)CC(=O)O.Fc1ccc(cc1)c2[n](c(c(c2c4ccccc4)\C(=N\c3ccccc3)\[O-])C(C)C)CC[C@@H](O)C[C@@H](O)CC(=O)[O-].O.O.O

InChI

1S/2C33H35FN2O5.Ca.3H2O/c2*1-21(2)31-30(33(41)35-25-11-7-4-8-12-25)29(22-9-5-3-6-10-22)32(23-13-15-24(34)16-14-23)36(31)18-17-26(37)19-27(38)20-28(39)40;;;;/h2*3-16,21,26-27,37-38H,17-20H2,1-2H3,(H,35,41)(H,39,40);;3*1H2/q;;+2;;;/p-2/t2*26-,27-;;;;/m11..../s1

InChI key

SHZPNDRIDUBNMH-NIJVSVLQSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Atorvastatin Calcium is a synthetic selective inhibitor of HMG-CoA reductase. It is insoluble in aqueous solution with pH below 4 and is freely soluble in methanol.

Application

Atorvastatin calcium USP reference standard intended for use in specified quality tests and assays.

Also used to prepare system suitability, standard, system suitability stock, identification, and standard stock solution for assay and impurity analysis according to the given below monographs of United States Pharmacopeia (USP):
  • Atorvastatin Calcium
  • Atorvastatin Calcium Tablets
  • Amlodipine and Atorvastatin Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Atorvastatin Calcium
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 35(1), 414-414 (2022)
Atorvastatin and Atorvastatin Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(2), 265-265 (2020)
Atorvastatin Calcium Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(1), 418-418 (2020)
Peter Norsk et al.
The Journal of physiology, 593(3), 573-584 (2015-03-17)
Weightlessness in space induces initially an increase in stroke volume and cardiac output, accompanied by unchanged or slightly reduced blood pressure.It is unclear whether these changes persist throughout months of flight.Here, we show that cardiac output and stroke volume increase
Xiao-ling Li et al.
International immunopharmacology, 23(2), 546-549 (2014-12-03)
This study aimed to investigate the efficacy of combined atorvastatin calcium and methylprednisolone for the treatment of multiple sclerosis relapse. Patients with multiple sclerosis (MS) at the relapse phase were randomized to receive either combined treatment of atorvastatin calcium and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service